siRNA Details
Gene Symbol: PCDHB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018940.3 | NP_061763.1 | 1 | 347 | ||
GenBank mRNA | AF152500.1 | - | 1 | 132 | ||
AF217750.1 | - | 1 | 132 | |||
AK222678.1 | - | 1 | 292 | |||
AK315054.1 | - | 1 | 350 | |||
BC130331.1 | - | 1 | 214 | 40146362 | ||
BC130333.1 | - | 1 | 214 | 40146364 | ||
DA779188.1 | - | 1 | 348 |
