siRNA Details
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018961.3 | NP_061834.1 | 5 | 706 | ||
XM_006724013.3 | XP_006724076.1 | 5 | 790 | |||
XM_011529605.2 | XP_011527907.1 | 5 | 706 | |||
XM_011529606.2 | XP_011527908.1 | 5 | 675 | |||
XM_011529607.2 | XP_011527909.1 | 5 | 759 | |||
XM_011529609.1 | XP_011527911.1 | 5 | 706 | |||
GenBank mRNA | AF520809.1 | - | 5 | 633 | ||
AJ277750.1 | - | 5 | 653 | |||
BC069357.1 | - | 5 | 653 | 7262315 | ||
BC069483.1 | - | 5 | 653 | 7262391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
