siRNA Details
Gene Symbol: TMEM238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190764.1 | NP_001177693.1 | 2 | 645 | ||
GenBank mRNA | BC052596.1 | - | 2 | 621 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190764.1 | NP_001177693.1 | 2 | 645 | ||
GenBank mRNA | BC052596.1 | - | 2 | 621 |