siRNA Details
Gene Symbol: TMEM238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190764.1 | NP_001177693.1 | 1 | 278 | ||
GenBank mRNA | BC052596.1 | - | 1 | 254 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029384, BC132101, BC144965.
