siRNA Details
Gene Symbol: HCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002139.2 | 1 | 415 | |||
GenBank mRNA | AF036977.1 | - | 1 | 415 | ||
BC011879.1 | - | 1 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
