siRNA Details
Gene Symbol: C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199534.1 | NP_001186463.1 | 2, 3 | 443 | ||
NM_018840.4 | NP_061328.1 | 2, 3 | 443 | |||
GenBank mRNA | AF112213.1 | - | 2, 3 | 407 | ||
AF274936.1 | - | 2, 3 | 338 | |||
AF274948.1 | - | 2, 3 | 256 | |||
BC001871.1 | - | 2, 3 | 360 | 3546888 | ||
BI223439.1 | - | 2, 3 | 448 | |||
BU848918.1 | - | 2, 3 | 402 | |||
S83364.1 | - | 2, 3 | 183 |
Gene Symbol: TGIF2-C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199535.1 | NP_001186464.1 | 3, 4 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
