siRNA Details
Gene Symbol: C6orf132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164446.2 | NP_001157918.1 | 4 | 3443 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C6orf132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164446.2 | NP_001157918.1 | 4 | 3443 |