siRNA Details
Gene Symbol: LOC390877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026804.1 | XP_016882293.1 | 2 | 329 | ||
XM_017026805.1 | XP_016882294.1 | 3 | 796 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC390877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026804.1 | XP_016882293.1 | 2 | 329 | ||
XM_017026805.1 | XP_016882294.1 | 3 | 796 |