siRNA Details
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318865.1 | NP_001305794.1 | 6 | 800 | ||
NM_018960.5 | NP_061833.1 | 6 | 857 | |||
NR_134899.1 | 5 | 714 | ||||
GenBank mRNA | AF101477.1 | - | 6 | 853 | ||
BC032627.1 | - | 6 | 849 | 5229272 | ||
FJ224320.1 | - | 6 | 853 | |||
KU178603.1 | - | 6 | 843 | |||
KU178604.1 | - | 6 | 786 | |||
KU178605.1 | - | 5 | 700 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 6 | 1229 | ||
NM_001318857.1 | NP_001305786.1 | 5 | 1101 | |||
NM_001318858.1 | NP_001305787.1 | 5 | 993 | |||
NR_134890.1 | 5 | 1055 | ||||
NR_134891.1 | 4 | 958 | ||||
NR_134892.1 | 5 | 1086 | ||||
GenBank mRNA | BQ947041.1 | - | 6 | 862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
