siRNA Details
Gene Symbol: ANKDD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276713.1 | NP_001263642.1 | 6 | 842 | ||
XM_011543618.2 | XP_011541920.1 | 6 | 1029 | |||
XM_011543623.2 | XP_011541925.1 | 2 | 594 | |||
XM_017009814.1 | XP_016865303.1 | 5 | 1134 | |||
XM_017009815.1 | XP_016865304.1 | 5 | 1134 | |||
XM_017009816.1 | XP_016865305.1 | 5 | 1134 |
