siRNA Details
Gene Symbol: LOC283911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833480.1 | - | 1 | 5067 |
Gene Symbol: FAM85B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CD106111.1 | - | 4 | 774 |
Gene Symbol: LOC100506990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040092.1 | 6 | 1698 | |||
GenBank mRNA | AK094417.1 | - | 2 | 1068 | ||
AK095981.1 | - | 6 | 1348 | |||
AK129982.1 | - | 7 | 1834 | |||
AL137298.1 | - | 4 | 789 |
