siRNA Details
Gene Symbol: C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199534.1 | NP_001186463.1 | 3 | 545 | ||
NM_018840.4 | NP_061328.1 | 3 | 545 | |||
NR_026562.3 | 4 | 656 | ||||
GenBank mRNA | AF112213.1 | - | 3 | 509 | ||
AF274936.1 | - | 3 | 443 | |||
AF274948.1 | - | 3 | 358 | |||
BC001871.1 | - | 3 | 462 | 3546888 | ||
BC004446.1 | - | 4 | 581 | 2819807 | ||
BU848918.1 | - | 3 | 504 |
Gene Symbol: TGIF2-C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199535.1 | NP_001186464.1 | 4 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
