siRNA Details
Gene Symbol: FLJ41278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033988.1 | 3 | 361 | |||
GenBank mRNA | AK123272.1 | - | 3 | 362 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ41278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033988.1 | 3 | 361 | |||
GenBank mRNA | AK123272.1 | - | 3 | 362 |