siRNA Details
Gene Symbol: LINC00675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036581.1 | 1 | 357 | |||
GenBank mRNA | AK024897.1 | - | 1 | 322 | ||
BC015790.1 | - | 1 | 295 | |||
BC041634.1 | - | 1 | 295 | |||
BM924455.1 | - | 1 | 310 | |||
DA425830.1 | - | 1 | 357 |
