siRNA Details
Gene Symbol: LINC01344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104175.1 | 3 | 541 | |||
GenBank mRNA | AK128015.1 | - | 5 | 486 | ||
BX096536.1 | - | 3 | 541 |
Gene Symbol: ZNF648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000302.1 | XP_016855791.1 | 1 | 1356 |
