siRNA Details
Gene Symbol: FAM182B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026714.2 | 4 | 2232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM182B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026714.2 | 4 | 2232 |