siRNA Details
Gene Symbol: C3orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168214.1 | NP_001161686.1 | 1 | 826 | ||
GenBank mRNA | BC031660.1 | - | 1 | 620 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C3orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168214.1 | NP_001161686.1 | 1 | 826 | ||
GenBank mRNA | BC031660.1 | - | 1 | 620 |