siRNA Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316897.1 | NP_001303826.1 | 4 | 948 | ||
NM_001316898.1 | NP_001303827.1 | 4 | 941 | |||
NR_133631.1 | 3 | 738 | ||||
GenBank mRNA | AK075259.1 | - | 4 | 941 | ||
AK130389.1 | - | 1 | 1021 | |||
BC012940.1 | - | 4 | 898 | 4478288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
