siRNA Details
Gene Symbol: EBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036592.1 | 2 | 665 | |||
GenBank mRNA | AK023696.1 | - | 1 | 1702 | ||
AK091718.1 | - | 2 | 665 | |||
BC032471.1 | - | 2 | 564 | |||
BC033624.1 | - | 2 | 564 | |||
BC080614.1 | - | 2 | 535 |
