siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 1 | 595 | |||
GenBank mRNA | AK024177.1 | - | 1 | 148 | ||
AK092686.1 | - | 1 | 53 | |||
BC038559.1 | - | 2 | 229 | |||
BC044229.1 | - | 1 | 155 | |||
DR006520.1 | - | 1 | 601 |
