siRNA Details
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 9 | 1267 | |||
GenBank mRNA | AL137492.1 | - | 4 | 438 | ||
BC030509.1 | - | 5 | 544 | |||
BC108680.1 | - | 8 | 1010 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 10 | 1248 | |||
GenBank mRNA | BC119762.1 | - | 6 | 693 |
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040584.2 | 8 | 992 | |||
GenBank mRNA | BC002581.2 | - | 9 | 942 | ||
BC119761.1 | - | 6 | 693 | 40119328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
