siRNA Details
Gene Symbol: C10orf126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278522.1 | NP_001265451.1 | 5 | 1740 | ||
XM_017016109.1 | XP_016871598.1 | 4 | 1656 | |||
GenBank mRNA | BC033839.1 | - | 5 | 1751 |
