siRNA Details
Gene Symbol: CISTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104333.1 | 1, 2 | 70 | |||
GenBank mRNA | DA125942.1 | - | 1, 2 | 70 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CISTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104333.1 | 1, 2 | 70 | |||
GenBank mRNA | DA125942.1 | - | 1, 2 | 70 |