siRNA Details
Gene Symbol: CISTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104332.1 | 3 | 609 | |||
NR_104333.1 | 2 | 334 | ||||
GenBank mRNA | DA125942.1 | - | 2 | 334 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CISTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104332.1 | 3 | 609 | |||
NR_104333.1 | 2 | 334 | ||||
GenBank mRNA | DA125942.1 | - | 2 | 334 |