siRNA Details
Gene Symbol: ZNF891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277291.1 | NP_001264220.1 | 2 | 1989 | ||
XM_017018666.1 | XP_016874155.1 | 2 | 2157 | |||
XM_017018667.1 | XP_016874156.1 | 2 | 1827 | |||
XM_017018668.1 | XP_016874157.1 | 2 | 1910 |
