siRNA Details
Gene Symbol: PFKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002021.2 | NP_001002021.2 | 4 | 587 | ||
XM_006724011.1 | XP_006724074.1 | 4 | 533 | |||
XM_006724012.1 | XP_006724075.1 | 2 | 318 | |||
XM_017028368.1 | XP_016883857.1 | 3 | 497 | |||
XM_017028369.1 | XP_016883858.1 | 3 | 337 | |||
GenBank mRNA | AK303605.1 | - | 3 | 436 | ||
BC006422.1 | - | 4 | 533 | 4040291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
