siRNA Details
Gene Symbol: LINC01136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034151.1 | 2 | 510 | |||
GenBank mRNA | AK309664.1 | - | 2 | 494 | ||
DA707241.1 | - | 2 | 510 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034151.1 | 2 | 510 | |||
GenBank mRNA | AK309664.1 | - | 2 | 494 | ||
DA707241.1 | - | 2 | 510 |