siRNA Details
Gene Symbol: LINC01272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278655.1 | NP_001265584.1 | 3 | 361 | ||
XM_006723680.3 | XP_006723743.1 | 3 | 374 | |||
GenBank mRNA | AK307192.1 | - | 3 | 349 | ||
BQ430993.1 | - | 3 | 361 |
