siRNA Details
Gene Symbol: C2orf81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145054.1 | NP_001138526.1 | 3 | 650 | ||
NM_001316764.1 | NP_001303693.1 | 3 | 402 | |||
NM_001316765.1 | NP_001303694.1 | 3 | 255 | |||
NM_001316766.1 | NP_001303695.1 | 2 | 485 | |||
GenBank mRNA | BU184705.1 | - | 2 | 446 | ||
HY047622.1 | - | 3 | 256 | |||
HY050307.1 | - | 3 | 404 |
