siRNA Details
Gene Symbol: THEGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256475.1 | NP_001243404.1 | 7 | 1059 | ||
XM_011534357.1 | XP_011532659.1 | 7 | 1093 | |||
XM_011534361.2 | XP_011532663.1 | 7 | 1093 | |||
XM_011534363.2 | XP_011532665.1 | 7 | 1093 | |||
XM_011534364.1 | XP_011532666.1 | 7 | 1093 | |||
XM_011534365.2 | XP_011532667.1 | 7 | 1093 | |||
GenBank mRNA | DY655427.1 | - | 2 | 51 | ||
DY655539.1 | - | 3 | 282 |
