siRNA Details
Gene Symbol: SPIN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019003.3 | NP_061876.3 | 2 | 604 | ||
XM_005262016.4 | XP_005262073.3 | 2 | 257 | |||
XM_005262017.4 | XP_005262074.3 | 2 | 257 | |||
XM_005262019.3 | XP_005262076.1 | 2 | 1719 | |||
XM_017029598.1 | XP_016885087.1 | 2 | 1664 | |||
GenBank mRNA | BC071694.1 | - | 2 | 604 | 5201217 | |
Y09858.1 | - | 1 | 648 |
Gene Symbol: SPIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006681.1 | NP_001006682.1 | 2 | 544 | ||
NM_001006682.1 | NP_001006683.1 | 2 | 492 | |||
NM_001006683.1 | NP_001006684.1 | 2 | 489 | |||
XM_005262008.4 | XP_005262065.2 | 2 | 338 | |||
XM_005262009.4 | XP_005262066.2 | 2 | 327 | |||
XM_005262010.1 | XP_005262067.1 | 2 | 549 | |||
XM_011530788.2 | XP_011529090.1 | 2 | 339 | |||
XM_017029552.1 | XP_016885041.1 | 2 | 336 | |||
GenBank mRNA | AF356353.1 | - | 2 | 361 | ||
BC000044.2 | - | 2 | 437 | 3504291 | ||
BC071974.1 | - | 2 | 476 | 6729986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
