siRNA Details
Gene Symbol: ATP8B5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003582.1 | 11 | 1604 | |||
NR_110320.1 | 11 | 1600 | ||||
GenBank mRNA | BC035229.1 | - | 10 | 3010 | ||
BC130448.1 | - | 6 | 1364 | |||
BC130450.1 | - | 6 | 1211 |
Gene Symbol: ZFAND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242911.1 | NP_001229840.1 | 5 | 725 | ||
NM_001242912.1 | NP_001229841.1 | 6 | 804 | |||
NM_001242913.1 | NP_001229842.1 | 7 | 759 | |||
NM_001242914.1 | NP_001229843.1 | 6 | 734 | |||
NM_001242915.1 | NP_001229844.1 | 5 | 570 | |||
NM_001242916.1 | NP_001229845.1 | 6 | 733 | |||
NM_001242917.1 | NP_001229846.1 | 6 | 733 | |||
NM_001242919.1 | NP_001229848.1 | 5 | 565 | |||
NM_019006.3 | NP_061879.2 | 6 | 888 | |||
XM_011521684.2 | XP_011519986.1 | 7 | 771 | |||
GenBank mRNA | AF061739.1 | - | 6 | 673 | ||
AF261138.1 | - | 5 | 559 | |||
AJ251095.1 | - | 5 | 522 | |||
AK055881.1 | - | 1 | 1682 | |||
AL136598.1 | - | 5 | 580 | |||
BC005283.1 | - | 6 | 777 | 4042180 | ||
BG720174.1 | - | 6 | 731 | |||
BP234517.1 | - | 5 | 514 | |||
CN428457.1 | - | 7 | 759 | |||
CR533476.1 | - | 4 | 466 | |||
DR762914.1 | - | 6 | 718 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_022985, AK002775, BC010683, AK144902, AJ251508.
Gene Ontology Categories:
