siRNA Details
Gene Symbol: CTC-338M12.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109909.1 | 1 | 61 | |||
GenBank mRNA | CF126280.1 | - | 1 | 61 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CTC-338M12.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109909.1 | 1 | 61 | |||
GenBank mRNA | CF126280.1 | - | 1 | 61 |