siRNA Details
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271844.1 | NP_001258773.1 | 8 | 5487 | ||
NM_001271845.1 | NP_001258774.1 | 8 | 5526 | |||
NM_019020.3 | NP_061893.2 | 12 | 6642 | |||
XM_005257049.2 | XP_005257106.1 | 12 | 6653 | |||
XM_005257050.3 | XP_005257107.1 | 12 | 6656 | |||
XM_006721694.3 | XP_006721757.1 | 13 | 6495 | |||
XM_011524322.2 | XP_011522624.1 | 12 | 6617 | |||
XM_011524323.2 | XP_011522625.2 | 10 | 6567 | |||
XM_011524324.2 | XP_011522626.1 | 8 | 6111 | |||
XM_017024186.1 | XP_016879675.1 | 10 | 6394 | |||
XM_017024187.1 | XP_016879676.1 | 11 | 6634 | |||
XM_017024188.1 | XP_016879677.1 | 10 | 5774 | |||
GenBank mRNA | AK000755.1 | - | 1 | 1416 | ||
AL157485.1 | - | 1 | 2248 | |||
AL512709.1 | - | 1 | 1882 | |||
BC080571.1 | - | 1 | 1431 | |||
CR936670.1 | - | 8 | 5542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
