siRNA Details
Gene Symbol: MIR124-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034102.2 | 2 | 532 | |||
NR_034103.2 | 2 | 353 | ||||
NR_109792.1 | 2 | 422 | ||||
NR_109793.1 | 2 | 394 | ||||
GenBank mRNA | AK124256.1 | - | 2 | 423 | ||
BM706891.1 | - | 1 | 335 | |||
BM717762.1 | - | 2 | 353 | |||
BQ639660.1 | - | 2 | 394 | |||
BX537900.1 | - | 1 | 314 | |||
DB176551.1 | - | 2 | 423 |
