siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 5 | 1920 | |||
GenBank mRNA | AK024177.1 | - | 5 | 1472 | ||
BC112296.1 | - | 3 | 583 | |||
BC112298.1 | - | 3 | 583 | |||
BG192193.1 | - | 1 | 59 |
