siRNA Details
Gene Symbol: LOC100132304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120493.1 | 3 | 438 | |||
NR_120494.1 | 3 | 426 | ||||
GenBank mRNA | HY042459.1 | - | 3 | 384 |
Gene Symbol: LINC00684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034116.1 | 3 | 438 | |||
NR_120499.1 | 3 | 426 | ||||
GenBank mRNA | AA393776.1 | - | 3 | 395 | ||
HY038238.1 | - | 3 | 384 |
