siRNA Details
Gene Symbol: FAM212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019099.4 | NP_061972.1 | 2 | 280 | ||
NM_198926.2 | NP_945120.1 | 2 | 138 | |||
XM_011541783.2 | XP_011540085.1 | 4 | 376 | |||
GenBank mRNA | AK055667.1 | - | 2 | 256 | ||
AK293989.1 | - | 2 | 165 | |||
AK295012.1 | - | 2 | 219 | |||
AL137198.1 | - | 2 | 187 | |||
AL137199.1 | - | 2 | 138 | |||
BC031558.1 | - | 2 | 235 | 5181834 | ||
DA306915.1 | - | 2 | 280 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175398, NM_001163356, AK078368, AK033558, BC052371.
