siRNA Details
Gene Symbol: MIOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019005.3 | NP_061878.3 | 12 | 2755 | ||
XM_005249780.3 | XP_005249837.1 | 11 | 2783 | |||
XM_005249781.3 | XP_005249838.1 | 10 | 2599 | |||
XM_005249782.3 | XP_005249839.1 | 10 | 2984 | |||
XM_011515432.2 | XP_011513734.1 | 8 | 2269 | |||
XM_017012361.1 | XP_016867850.1 | 11 | 2950 | |||
XM_017012362.1 | XP_016867851.1 | 11 | 2789 | |||
GenBank mRNA | AK000330.1 | - | 11 | 2624 | ||
AK096829.1 | - | 1 | 871 | |||
AK294561.1 | - | 12 | 2714 | |||
AL136892.1 | - | 7 | 1218 | |||
BC005883.2 | - | 9 | 1434 | |||
BC140833.1 | - | 11 | 2605 | 9021701 | ||
BC140834.1 | - | 10 | 2507 | 9021702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
