siRNA Details
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 7 | 2654 | ||
NM_019008.5 | NP_061881.2 | 6 | 3709 | |||
NR_130789.1 | 6 | 3623 | ||||
XM_011530236.1 | XP_011528538.1 | 8 | 3777 | |||
XM_011530237.1 | XP_011528539.1 | 7 | 3897 | |||
XM_011530238.1 | XP_011528540.1 | 7 | 3981 | |||
XM_017028838.1 | XP_016884327.1 | 7 | 3875 | |||
GenBank mRNA | AK000239.1 | - | 3 | 2880 | ||
AL365515.1 | - | 6 | 3586 | |||
AL834205.1 | - | 6 | 3551 | |||
BC002587.2 | - | 2 | 2851 | |||
BC008327.1 | - | 6 | 3563 | 3502711 |
