siRNA Details
Gene Symbol: RNF216P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015449.1 | 4, 5 | 590 | |||
GenBank mRNA | BC026736.1 | - | 4, 5 | 469 | ||
BX538059.1 | - | 4, 5 | 517 |
Gene Symbol: RNF216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY062174.1 | - | 4, 5 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
