siRNA Details
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271844.1 | NP_001258773.1 | 8 | 6201 | ||
NM_001271845.1 | NP_001258774.1 | 8 | 6240 | |||
NM_019020.3 | NP_061893.2 | 12 | 7356 | |||
XM_005257049.2 | XP_005257106.1 | 12 | 7367 | |||
XM_005257050.3 | XP_005257107.1 | 12 | 7370 | |||
XM_006721694.3 | XP_006721757.1 | 13 | 7209 | |||
XM_011524322.2 | XP_011522624.1 | 12 | 7331 | |||
XM_011524323.2 | XP_011522625.2 | 10 | 7281 | |||
XM_011524324.2 | XP_011522626.1 | 8 | 6825 | |||
XM_017024186.1 | XP_016879675.1 | 10 | 7108 | |||
XM_017024187.1 | XP_016879676.1 | 11 | 7348 | |||
XM_017024188.1 | XP_016879677.1 | 10 | 6488 | |||
GenBank mRNA | AK000755.1 | - | 1 | 2130 | ||
AL157485.1 | - | 1 | 2962 | |||
AL512709.1 | - | 1 | 2596 | |||
BC080571.1 | - | 1 | 2145 | |||
CR936670.1 | - | 8 | 6256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
