siRNA Details
Gene Symbol: KIAA1462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020848.2 | NP_065899.1 | 3 | 643 | ||
XM_011519608.1 | XP_011517910.1 | 3 | 682 | |||
XM_011519609.2 | XP_011517911.1 | 4 | 710 | |||
GenBank mRNA | AB040895.2 | - | 2 | 536 | ||
AK308850.1 | - | 3 | 625 | |||
BC153865.1 | - | 2 | 446 | |||
BX648192.1 | - | 4 | 732 |
