siRNA Details
Gene Symbol: KIAA1671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145206.1 | NP_001138678.1 | 7 | 5085 | ||
XM_005261793.2 | XP_005261850.1 | 9 | 5398 | |||
XM_005261794.4 | XP_005261851.1 | 9 | 5351 | |||
XM_006724346.2 | XP_006724409.1 | 10 | 5337 | |||
GenBank mRNA | AB051458.1 | - | 4 | 733 | ||
AL832019.1 | - | 4 | 4467 | |||
BC171801.1 | - | 7 | 5085 | 9054455 |
