siRNA Details
Gene Symbol: THAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130475.2 | NP_001123947.1 | 3 | 3036 | ||
NM_001287598.1 | NP_001274527.1 | 3 | 2990 | |||
NM_001287599.1 | NP_001274528.1 | 3 | 2914 | |||
NM_001287601.1 | NP_001274530.1 | 2 | 2843 | |||
NM_182529.3 | NP_872335.2 | 3 | 3112 | |||
GenBank mRNA | AL833137.1 | - | 3 | 2970 |
