siRNA Details
Gene Symbol: ING3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019071.2 | NP_061944.2 | 12 | 1721 | ||
GenBank mRNA | AF074968.1 | - | 12 | 1698 | ||
AK000096.1 | - | 12 | 1722 | |||
AL603623.1 | - | 10 | 5506 | |||
AY007790.1 | - | 12 | 1702 | |||
BC010851.1 | - | 11 | 1571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
