siRNA Details
Gene Symbol: L1TD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164835.1 | NP_001158307.1 | 4 | 1419 | ||
NM_019079.4 | NP_061952.3 | 3 | 1308 | |||
GenBank mRNA | AK001746.1 | - | 1 | 677 | ||
AK001973.1 | - | 3 | 1234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
