siRNA Details
Gene Symbol: LCE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178428.3 | NP_848515.1 | 2 | 580 | ||
GenBank mRNA | BG217525.1 | - | 1 | 554 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LCE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178428.3 | NP_848515.1 | 2 | 580 | ||
GenBank mRNA | BG217525.1 | - | 1 | 554 |