siRNA Details
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | J00194.1 | - | 4, 5 | 810 | ||
K01171.1 | - | 4, 5 | 798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
