siRNA Details
Gene Symbol: KRTAP19-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303120.1 | NP_001290049.1 | 1 | 190 | ||
GenBank mRNA | AB096947.1 | - | 1 | 188 | ||
AB096948.1 | - | 1 | 188 | |||
BC101813.1 | - | 1 | 190 | |||
BC101815.1 | - | 1 | 190 |
